Learn More
Hach Company sensION+ 5055 portable ORP electrode for general purpose applications
Supplier:  Hach Company LZW5055.97.0002
Encompass_Preferred
sensION+ 5055 is a combination platinum ORP electrode with a polycarbonate body and non-refillable gel-electrolyte reference. The 5055 has a fixed 1 meter cable and MP5 connector dedicated for use with Hach sensION+ Portable pH (mV) meters. The 5055 is ideal for oxidation-reduction potential (ORP) measurements in general aqueous applications.
Catalog No. 50-199-7971
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.