Learn More
Hach Company Sension+ 5049 portable multi-parameter electrode: pH, conductivity, DO, and temperature
Supplier:  Hach Company LZW5049.97.0002
Encompass_Preferred
Sension+ 5049 is a multi-parameter electrode for determination of pH, conductivity, DO and temperature. The 5049 has a plastic body, and 3 parameter-specific, separately attached electrodes. The probe has a fixed 1 meter cable and MP8 connector dedicated for use with Hach Sension+ MM156 portable meters. It is ideal for pH, conductivity, and DO measurements in general aqueous applications.
Catalog No. 50-199-7967
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.