Learn More
Hach Company Sension+ 5021T laboratory combination pH electrode for "difficult" (LIS) applications
Supplier:  Hach Company LZW5021T.97.002
Encompass_Preferred
Sension+ 5021T is a glass combination pH electrode with a refillable reference electrolyte and built-in temperature sensor. The 5021T has a fixed 1 meter cable with BNC connector (pH) and banana (temperature) connectors. It is intended for use with Hach Sension+ Laboratory pH meters. The 5021T has sleeve junction and an encapsulated reference system with silver ion barrier. It and is ideal for pH measurements in low ionic strength applications, samples with colloids, wines and paints or viscous samples such as emulsions and creams.
Catalog No. 50-199-7963
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.