Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
502.2 VOA Calibration Kit #2 (2000ppm), Restek™
Shop All Restek Corporation Products
Click to view available options
Quantity:
1 mL
Description
Contains 1mL each of these mixtures.
30042: 502.2 Calibration Mix 1 (gases)
- Bromomethane (74-83-9)
- Chloroethane (75-00-3)
- Chloromethane (74-87-3)
- Dichlorodifluoromethane (CFC-12) (75-71-8)
- Trichlorofluoromethane (CFC-11) (75-69-4)
- Vinyl chloride (75-01-4)
30431: 502.2 MegaMix Standard
- Benzene (71-43-2)
- Bromobenzene (108-86-1)
- Bromochloromethane (74-97-5)
- Bromodichloromethane (75-27-4)
- Bromoform (75-25-2)
- n-Butylbenzene (104-51-8)
- sec-Butylbenzene (135-98-8)
- tert-Butylbenzene (98-06-6)
- Carbon tetrachloride (56-23-5)
- Chlorobenzene (108-90-7)
- Chloroform (67-66-3)
- 2-Chlorotoluene (95-49-8)
- 4-Chlorotoluene (106-43-4)
- Dibromochloromethane (124-48-1)
- 1,2-Dibromo-3-chloropropane (DBCP) (96-12-8)
- 1,2-Dibromoethane (EDB) (106-93-4)
- Dibromomethane (74-95-3)
- 1,2-Dichlorobenzene (95-50-1)
- 1,3-Dichlorobenzene (541-73-1)
- 1,4-Dichlorobenzene (106-46-7)
- 1,1-Dichloroethane (75-34-3)
- 1,2-Dichloroethane (107-06-2)
- 1,1-Dichloroethene (75-35-4)
- cis-1,2-Dichloroethene (156-59-2)
- trans-1,2-Dichloroethene (156-60-5)
- 1,2-Dichloropropane (78-87-5)
- 1,3-Dichloropropane (142-28-9)
- 2,2-Dichloropropane (594-20-7)
- 1,1-Dichloropropene (563-58-6)
- cis-1,3-Dichloropropene (10061-01-5)
- trans-1,3-Dichloropropene (10061-02-6)
- Ethylbenzene (100-41-4)
- Hexachloro-1,3-butadiene (hexachlorobutadiene) (87-68-3)
- Isopropylbenzene (cumene) (98-82-8)
- 4-Isopropyltoluene (p-cymene) (99-87-6)
- Methylene chloride (dichloromethane) (75-09-2)
- Naphthalene (91-20-3)
- n-Propylbenzene (103-65-1)
- Styrene (100-42-5)
- 1,1,1,2-Tetrachloroethane (630-20-6)
- 1,1,2,2-Tetrachloroethane (79-34-5)
- Tetrachloroethene (127-18-4)
- Toluene (108-88-3)
- 1,2,3-Trichlorobenzene (87-61-6)
- 1,2,4-Trichlorobenzene (120-82-1)
- 1,1,1-Trichloroethane (71-55-6)
- 1,1,2-Trichloroethane (79-00-5)
- Trichloroethene (79-01-6)
- 1,2,3-Trichloropropane (96-18-4)
- 1,2,4-Trimethylbenzene (95-63-6)
- 1,3,5-Trimethylbenzene (108-67-8)
- m-Xylene (108-38-3)
- o-Xylene (95-47-6)
- p-Xylene (106-42-3)
Specifications
Specifications
Concentration | 2000ppm |
Quantity | 1 mL |
Type | Multi-component Standard |
Safety and Handling
ShelfLife | 6 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction