Learn More
Hach Company Sension+ 5011T laboratory combination pH electrode for "dirty" (wastewater) applications
Supplier:  Hach Company LZW5011T.97.002
Encompass_Preferred
Sension+ 5011T is a glass combination pH electrode with a non-refillable solid polymer electrolyte reference and built-in temperature sensor. The 5011T has a fixed 1 meter cable with BNC connector (pH) and banana (temperature) connectors. It is intended for use with Hach Sension+ Laboratory pH meters. The 5011T has an open annular junction and is ideal for pH measurements in wastewater, foods, and other “dirty” samples with high solids content.
Catalog No. 50-199-7960
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.