Learn More
MTC Bio Centrifuge Tube, 500mL, PP, sterile, with screw-cap, 8 per bag, 5 bags per case, 40/cs
Supplier:  MTC Bio C1500
Encompass_Preferred
500mL screw-cap polypropylene tubes are manufactured of polypropylene for temperature and chemical resistance. These tubes are intended for large volume centrifugation and are widely used in cell harvesting/purification and drug discovery applications. MTC Bio's 500mL centrifuge tubes can withstand up to 6,000xg maximum RCF*. They are supplied in sterile, RNase, DNase, and pyrogen-free packs of 5 tubes, 40 tubes per case.
Catalog No. 50-226-8451
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.