Learn More
Chemglass Life Sciences #50 Internal Thread, Glass Only
SDP
Supplier:  Chemglass Life Sciences CGB36006
Encompass_Preferred
#50 Internal Thread, Glass Only. 57mm Tube OD, Accepts 47-48mm OD Tubing. Internal glass thread for use with CG-362, CG-363, CG-364, and CG-366 PTFE bushings. Tubing OD listed is the shank OD of the thread.
Catalog No. 50-153-1988
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.