Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodo-2′-Deoxyuridine, MP Biomedicals™

Catalog No. MP21003572
Click to view available options
Quantity:
250 mg

Active against vaccinia virus (orthopoxviruses). 5-Iodo-2′-Deoxyuridine, MP Biomedicals is a pyrimidine analog (halogenated thymidine) that compete with thymidine for incorporation into DNA of replicating cells. This incorporation results in radio sensiti

Chemical Identifiers

CAS 54-42-2
Molecular Formula C9H11IN2O5
Molecular Weight (g/mol) 354.10
MDL Number MFCD00134656
InChI Key XQFRJNBWHJMXHO-RRKCRQDMSA-N
Synonym idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
PubChem CID 5905
ChEBI CHEBI:147675
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O

Specifications

Melting Point 196°C to 197°C (literature)
Density 2.148g/cm3 (Lit.)
Quantity 250 mg
Solubility Information Solubility at 25°C in mg/mL: 2.0 in water; 2.0 in 0.2N HCl; 74.0 in 0.2N NaOH; 4.4 in methanol; 2.6 in alc; 0.014 in ether; 0.003 in chloroform; 1.6 in acetone; 1.8 in ethyl acetate; 5.7 in dioxane.
Formula Weight 354.1
Percent Purity ∼99% by HPLC
Physical Form Powder
Recommended Storage Store at 0°C. Protect from light.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.