Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxy-2-methylbenzoic acid, 97%, Thermo Scientific™

Catalog No. AAH6077403
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Hydroxy-2-methylbenzoic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible wit

Chemical Identifiers

CAS 578-22-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD09833184
InChI Key ZIOYQUNKXJQXQY-UHFFFAOYSA-N
Synonym 5-hydroxy-2-methyl-benzoic acid, 5,2-cresotic acid, 5-hydroxy-o-toluic acid, benzoic acid, 5-hydroxy-2-methyl, 2-methyl-5-hydroxybenzoic acid, acmc-1b0c9, ksc494s9h, 5-hydroxy-2-methylbenzoic acid
PubChem CID 235188
IUPAC Name 5-hydroxy-2-methylbenzoic acid
SMILES CC1=CC=C(O)C=C1C(O)=O

Specifications

Melting Point 182°C to 187°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material 5-Hydroxy-2-methylbenzoic acid
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.