Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-methyl-1H-indene-3-acetic acid, 97%, Thermo Scientific™

Catalog No. AAH61410MD
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 32004-66-3
Molecular Formula C12H11FO2
Molecular Weight (g/mol) 206.216
MDL Number MFCD01076194
InChI Key QDDPPRDVFIJASZ-UHFFFAOYSA-N
Synonym 5-fluoro-2-methyl-1h-inden-3-yl acetic acid, 2-5-fluoro-2-methyl-1h-inden-3-yl acetic acid, 5-fluoro-2-methyl-1h-indene-3-acetic acid, 5-fluoro-2-methylindene-3-acetic acid, 5-fluoro-2-methyl-3-indenyl-acetic acid, 6-fluoro-2-methyl-3h-inden-1-yl acetic acid, 2-6-fluoro-2-methyl-3h-inden-1-yl acetic acid, fluoromethylindenylaceticacid, 5-fluoro-2-methyl-3-indenylacetic acid, 5-fluoro-2-methylinden-3-yl acetic acid
PubChem CID 3015608
IUPAC Name 2-(6-fluoro-2-methyl-3H-inden-1-yl)acetic acid
SMILES CC1=C(C2=C(C1)C=CC(=C2)F)CC(=O)O

Specifications

Quantity 250 mg
Formula Weight 206.21
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methyl-1H-indene-3-acetic acid
Hazard Category H300-H312+H332-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c
EINECSNumber 250-891-7
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.