Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-Chloro-3-Indolyl-β-D-Glucuronide Cyclohexylammonium Salt, MP Biomedicals™

Catalog No. MP219392480
Click to view available options
Quantity:
100 mg

Product is a β-glucuronidase substrate, which forms an intense blue precipitate upon enzymatic action. Product is used for the detection of the GUS gene in bacterial colonies and in histochemical applications.

Chemical Identifiers

CAS 18656-96-7
Molecular Formula C20H26BrClN2O7
Molecular Weight (g/mol) 521.789
InChI Key JXCKZXHCJOVIAV-CYRSAHDMSA-N
Synonym c14h13brclno7.c6h13n, cyclohexanaminium 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, 5-bromo-4-chloro-3-indolyl-beta-d-glucuronide cyclohexylammonium salt, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, 5-bromo-4-chloro-3-indolyl-beta-d-glucuronide cyclohexyl ammonium salt, 5-bromo-4-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt monohydrate
PubChem CID 42609655
IUPAC Name (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium
SMILES C1CCC(CC1)[NH3+].C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br

Specifications

Melting Point 241°C to 243°C (literature)
Quantity 100 mg
Solubility Information Soluble in DMF (2mg/mL; clear, colorless solution).
Optical Rotation (c=1, DMF : H2O 1:1): –90.4 db (Lit.)
Formula Weight 521.8g/mol
Percent Purity ≥95%
Physical Form Crystalline Powder
Recommended Storage Store at –20°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.