Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH59519MD
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1449135-41-4
Molecular Formula C17H25BFNO4
Molecular Weight (g/mol) 337.198
MDL Number MFCD22419260
InChI Key JWYMEKZRPHNWGS-UHFFFAOYSA-N
Synonym tert-butyl n-4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 5-boc-amino-2-fluorobenzeneboronic acid pinacol ester, tert-butyl 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate
PubChem CID 86810446
IUPAC Name tert-butyl N-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)NC(=O)OC(C)(C)C)F

Specifications

Melting Point 130°C to 132°C
Quantity 250 mg
Formula Weight 337.2
Percent Purity 96%
Chemical Name or Material 5-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.