Learn More
Andwin Scientific 4OZ CYTOSEAL LOW VISCOSIT
Supplier: Andwin Scientific 48212154
The Cytoseal Mounting Medium is a clear colorless medium with low viscosity. It eliminates the chance of cracking, fading, and discoloration of stained specimens with Thermo Scientific Richard-Allan Scientific Cytoseal 60 and 280 mounting media. This maintain the specimen's original quality almost indefinitely. This product is ideal for use with cover glass. It is sold individually in a quantity of 4 oz.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.