Learn More
Thomas Scientific Axygenr 48 Well Clear V-Bottom 5mL Polypropylene Rectangular Well Deep Well Plate, 5 per Pack, Nonsterile
Supplier: Thomas Scientific 1223T83
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C. Features 5.0mL deep well plate with rectangular wells ANSI/SLAS 1-2004: Microplates - Footprint Dimensions Ultra-flat surface ensures proper sealing with heat sealing film
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.