Learn More
Tocris Bioscience™ (RS)-3,5-DHPG
Selective group I mGlu agonist
$113.60 - $984.00
Specifications
Chemical Name or Material | (RS)-3,5-Dihydroxyphenylglycine |
---|---|
CAS | 19641-83-9 |
Assay Percent Range | >99% |
Solubility | Soluble to 10mM in water |
Molecular Formula | C8H9NO4 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
03-421-0
|
Tocris Bioscience™
0342/10 |
10 mg |
Each for $238.00
|
|
|||||
03-421-R
|
Tocris Bioscience™
0342/1 |
1 mg |
Each for $113.60
|
|
|||||
03-425-0
|
Tocris Bioscience™
0342/50 |
50 mg |
Each for $984.00
|
|
|||||
Description
Selective group I metabotropic glutamate receptor agonist which activates both mGlu1 and mGlu5. Also reported to be an antagonist at metabotropic glutamate receptors linked to phospholipase D.S-enantiomer also available.
Specifications
(RS)-3,5-Dihydroxyphenylglycine | |
>99% | |
C8H9NO4 |
19641-83-9 | |
Soluble to 10mM in water | |
Observed MW: 183.16 |
Safety and Handling
Recommended Storage : Desiccate at -20°C
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.