Learn More
Electron Microscopy Sciences Multi Holder 45 ° for 6 pin stubs, 3.2 mm x 8mm Pin
Supplier:  Electron Microscopy Sciences 7595270
Encompass_Preferred
This 35 mm in diameter 45 Degree multi pin holder is designed to save time. It allows for a higher SE signal without having to tilt the specimen. It is made from machined aluminum with stainless steel Allen set screws. It accommodates up to six standard 12.7 mm (1/2") pin stubs.
Type: 3.2 mm x 8 mm Pin
Catalog No. 50-190-1522
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.