Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)thiobenzamide, 97%, Thermo Scientific™

Catalog No. AAH5858503
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 149169-34-6
Molecular Formula C8H6F3NOS
Molecular Weight (g/mol) 221.197
MDL Number MFCD00068113
InChI Key GHQSKHMIUWHKHO-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy thiobenzamide, 4-trifluoromethoxy benzenecarbothioamide, benzenecarbothioamide,4-trifluoromethoxy, acmc-20e60i, 4-trifluoromethoxythiobenzamide, 4-trifluoromethoxy-thiobenzamide, 4-trifluoromethoxy benzothioamide, 4-trifluoromethyloxy benzenecarbothioamide, 4-trifluoromethoxy benzene-1-carbothioamide, amino 4-trifluoromethoxy phenyl methane-1-thione
PubChem CID 2779333
IUPAC Name 4-(trifluoromethoxy)benzenecarbothioamide
SMILES C1=CC(=CC=C1C(=S)N)OC(F)(F)F

Specifications

Melting Point 124°C to 126°C
Quantity 1 g
Formula Weight 221.19
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)thiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.