Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)-3′-methylazobenzene 98.0+%, TCI America™
SDP

Catalog No. M020725G
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 55-80-1
Molecular Formula C15H17N3
Molecular Weight (g/mol) 239.322
MDL Number MFCD00048014
InChI Key LVTFSVIRYMXRSR-UHFFFAOYSA-N
Synonym 3′C-Methyl-4-(dimethylamino)azobenzene
PubChem CID 5934
IUPAC Name N,N-dimethyl-4-[(3-methylphenyl)diazenyl]aniline
SMILES CC1=CC(=CC=C1)N=NC2=CC=C(C=C2)N(C)C

Specifications

Melting Point 120°C
Quantity 25 g
Formula Weight 239.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)-3′-methylazobenzene
RTECSNumber BX8250000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.