Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4′-ethoxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. C315325G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 58743-78-5
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD01218033
InChI Key VETJRGXWDLHERN-UHFFFAOYSA-N
Synonym 4'-ethoxy-1,1'-biphenyl-4-carbonitrile, 4-cyano-4'-ethoxybiphenyl, 4-ethoxy-4'-cyanobiphenyl, 4-4-ethoxyphenyl benzonitrile, 4-ethoxy-1,1'-biphenyl-4'-carbonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-ethoxy, 4'-ethoxy-4-cyanobiphenyl, 4-4-ethoxyphenyl benzenecarbonitrile, pubchem2524
PubChem CID 93893
IUPAC Name 4-(4-ethoxyphenyl)benzonitrile
SMILES CCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 102°C
Color White
Quantity 25 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-ethoxybiphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.