Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloropiperidine 97.0+%, TCI America™
SDP

Catalog No. C21825G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5382-18-3
Molecular Formula C5H10ClN
Molecular Weight (g/mol) 119.592
InChI Key GBPBXBUHZSOKTH-UHFFFAOYSA-N
PubChem CID 420932
IUPAC Name 4-chloropiperidine
SMILES C1CNCCC1Cl

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
UN Number 2920
Formula Weight 119.59
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chloropiperidine
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.