Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-iodophenol 98.0+%, TCI America™
SDP

Catalog No. C31721G
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 71643-66-8
Molecular Formula C6H4ClIO
Molecular Weight (g/mol) 254.451
MDL Number MFCD08235248
InChI Key JKPLMQJLGBBFLO-UHFFFAOYSA-N
PubChem CID 620234
IUPAC Name 4-chloro-2-iodophenol
SMILES C1=CC(=C(C=C1Cl)I)O

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 1 g
Formula Weight 254.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-iodophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.