Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals

Catalog No. AAA1214614
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 445-02-3
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
MDL Number MFCD00064393
InChI Key PHXGKHTWEOPCEW-UHFFFAOYSA-N
Synonym 2-amino-5-bromobenzotrifluoride, 4-bromo-2-trifluoromethyl aniline, benzenamine, 4-bromo-2-trifluoromethyl, 4-bromo-2-trifluoromethyl-phenylamine, 2-amino-5-bromo trifluoromethyl benzene, 2-amino-5-bromo benzotrifluoride, 4-bromo-2-trifluoroaniline, 2-trifluoromethyl-4-bromoaniline, 4-bromo-alpha,alpha,alpha-trifluoro-o-toluidine, 4-bromo-2-trifluoromethyl-aniline
PubChem CID 67960
IUPAC Name 4-bromo-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(Br)C=C1C(F)(F)F

Specifications

Density 1.71
Boiling Point 84°C to 85°C (5 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.532
Quantity 25 g
UN Number UN2810
Beilstein 2211504
Formula Weight 240.03
Percent Purity 98%
Chemical Name or Material 4-Bromo-2- (trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 207-150-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.