Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. AAH59944MD
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1326283-60-6
Molecular Formula C13H19BFNO3
Molecular Weight (g/mol) 267.11
MDL Number MFCD22419265
InChI Key JWACNMHKJHEWAQ-UHFFFAOYSA-N
Synonym 4-amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 5-fluoro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
PubChem CID 67363295
IUPAC Name 5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 250 mg
Formula Weight 267.11
Percent Purity 96%
Chemical Name or Material 4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.