Learn More
Sklar Instruments Scissors, Sklar Iris Scissors, Sklar 4 -1/2 in., Sharp/Sharp, Straight, Micro-Serrated, Stainless Steel, EA.
Supplier: Sklar Instruments 153585
Scissors, Sklarcut Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Micro-Serrated, Stainless Steel, EA, Sklarcut Iris scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Sklarcut scissors are identified by a single gold coated finger ring on the handle of the serrated blade.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.