Learn More
Sklar Instruments Scissors, Sklar Plastic Surgery Scissors, Sklar 4-1/2 in., Sharp/Sharp, Curved, Micro-Serrated, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 153525
Encompass_Preferred
Scissors, Sklarcut Plastic Surgery Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Micro-Serrated, Stainless Steel, EA. sklarcut Plastic Surgery Scissors are non-ratcheted, finger ring scissors used for cutting and dissecting delicate tisssue in various plastic surgery procedures. Sklarcut scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made. Sklarcut scissors are identified by a single gold coated finger ring on the handle of the serrated blade. This product is curved with sharp/sharp tips, and an overall length of 4-1/2 inches.
Catalog No. 50-109-3465
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.