Learn More
Sklar Instruments Scissors, McPherson Vannas Scissors, Sklar 3 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
Supplier: Sklar Instruments 641960
Scissors, McPherson Vannas Scissors, Sklar, 3 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Sklar's McPherson-Vannas Scissors are double spring thumb scissors commonly used in ophthalmic procedures such as iridotomy. These scissors feature 10mm, sharp, delicate blades ideal for use on fine tissues of the eye. The handle has a horizontally serrated grip that provides greater control of the instrument. This product has straight blades and an overall length of 3 inches.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.