Learn More
Epredia™ 3DHistech TMA Image Analysis Module Plug-in - 10 Users
Epredia™ 3DHistech TMA Image module is an application that enables researchers to deal with a large number of samples very easily in a structured way and makes the evaluation and scoring of several hundred samples possible.
Supplier:  Epredia™ 91024
Catalog No. 91-024
Explore all special offers
Add to Cart
Description
Description
  • Project-based workflow: link multiple blocks of multiple slides together in one project. This enables you to deal with huge number of samples very easily in a structured way. Slides can be added to and removed from an existing project.
  • Data of samples and blocks and also the map of blocks are presented in an .xls file.
  • Based on the map of blocks the program detects the cores and assigns the selected attributes to them.
  • Scoring of cores is easy and convenient: TMA is integrated with SlideViewer (former CaseViewer) so seamless zooming and navigation is available. Range of values of scoring can be set by the user. After scoring the virtual objective jumps to the next core so you don't have to find it. The scoring results are saved in the XLS database.
  • Simple (1-dimensional) and combined (2-dimensional) scoring schemes can be set by the user.
  • The cores can be filtered based on scores and other attributes, including measurement data created by the quantification software applications.
  • The data of scoring can be analyzed with third-party software.
  • Research use only
Specifications
Specifications
Image Analysis Module Plug In
Microarray Scanner
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.