Learn More
Chemplex Industries Mylar 3.6 μm (0.14 mil); 1.38"; SpectroMembrane Thin-Film Sample Supports
Supplier:  Chemplex Industries 301435
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Features: Thin-Film mounted on a carrier frame, Perforation Sizes, 1.35" (35.0mm), 1.77" (45.0mm), 2.17" (55.0mm), 2.50" (63.5mm), 3.00" (76.2mm), 100/pk
Catalog No. NC1839853