Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitro-N-phenylbenzamide, 97%, Thermo Scientific™

Catalog No. AAH60695MD
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2243-73-4
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD00017021
InChI Key VWIRWLAPFZXYSL-UHFFFAOYSA-N
Synonym 3-nitrobenzanilide, 3-nitro-n-phenyl-benzamide, benzamide, 3-nitro-n-phenyl, 3-nitrophenyl-n-benzamide, 3-nitro-n-phenyl benzamide
PubChem CID 347755
IUPAC Name 3-nitro-N-phenylbenzamide
SMILES C1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)[N+](=O)[O-]

Specifications

Quantity 250 mg
Formula Weight 242.23
Percent Purity 97%
Chemical Name or Material 3-Nitro-N-phenylbenzamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.