Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

$62.00

Chemical Identifiers

CAS 182344-13-4
Molecular Formula C6H6BClO3
Molecular Weight (g/mol) 172.37
MDL Number MFCD09258739
InChI Key WWQIKFZZILXJHG-UHFFFAOYSA-N
Synonym 3-chloro-4-hydroxyphenyl boronic acid, 3-chloro-4-hydroxybenzeneboronic acid, 4-borono-2-chlorophenol, boronic acid, 3-chloro-4-hydroxyphenyl, acmc-1bwc3, 3-chloro-4-hydroxyphenylboronicacid, 3-chloro-4-hydroxy-phenylboronic acid, 3-chloro-4-hydroxy-phenyl boronic acid, 3-chloranyl-4-oxidanyl-phenyl boronic acid
PubChem CID 22125373
IUPAC Name (3-chloro-4-hydroxyphenyl)boronic acid
SMILES OB(O)C1=CC(Cl)=C(O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
C29691G
SDP
View Documents
TCI America
C29691G
1 g
Each for $62.00
Add to Cart
 
C2969-5G
SDP
View Documents
TCI America
C29695G
5 g N/A N/A N/A
Description

Description

Specifications

Chemical Identifiers

182344-13-4
172.37
WWQIKFZZILXJHG-UHFFFAOYSA-N
22125373
OB(O)C1=CC(Cl)=C(O)C=C1
C6H6BClO3
MFCD09258739
3-chloro-4-hydroxyphenyl boronic acid, 3-chloro-4-hydroxybenzeneboronic acid, 4-borono-2-chlorophenol, boronic acid, 3-chloro-4-hydroxyphenyl, acmc-1bwc3, 3-chloro-4-hydroxyphenylboronicacid, 3-chloro-4-hydroxy-phenylboronic acid, 3-chloro-4-hydroxy-phenyl boronic acid, 3-chloranyl-4-oxidanyl-phenyl boronic acid
(3-chloro-4-hydroxyphenyl)boronic acid

Specifications

182344-13-4
MFCD09258739
WWQIKFZZILXJHG-UHFFFAOYSA-N
(3-chloro-4-hydroxyphenyl)boronic acid
22125373
Crystalline Powder
C6H6BClO3
3-chloro-4-hydroxyphenyl boronic acid, 3-chloro-4-hydroxybenzeneboronic acid, 4-borono-2-chlorophenol, boronic acid, 3-chloro-4-hydroxyphenyl, acmc-1bwc3, 3-chloro-4-hydroxyphenylboronicacid, 3-chloro-4-hydroxy-phenylboronic acid, 3-chloro-4-hydroxy-phenyl boronic acid, 3-chloranyl-4-oxidanyl-phenyl boronic acid
OB(O)C1=CC(Cl)=C(O)C=C1
172.37
172.37
3-Chloro-4-hydroxyphenylboronic Acid (contains varying amounts of Anhydride)
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.