Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-iodotoluene 95.0+%, TCI America™
SDP

Catalog No. C33645G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5100-98-1
Molecular Formula C7H6ClI
Molecular Weight (g/mol) 252.479
MDL Number MFCD00051756
InChI Key FTGLKPMFTLNUBN-UHFFFAOYSA-N
Synonym 3-chloro-2-iodotoluene, toluene, 3-chloro-2-iodo, benzene, 1-chloro-2-iodo-3-methyl, pubchem3655, acmc-1asrb, ksc493o0b, 1-chloro-2-iodo-3-methyl-benzene, buttpark 121\11-21, 1-chloranyl-2-iodanyl-3-methyl-benzene
PubChem CID 138414
IUPAC Name 1-chloro-2-iodo-3-methylbenzene
SMILES CC1=C(C(=CC=C1)Cl)I

Specifications

Boiling Point 125°C
Quantity 5 g
Formula Weight 252.48
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chloro-2-iodotoluene
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.