Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methoxybenzonitrile, 99%, Thermo Scientific Chemicals

Catalog No. AAA1669814
Click to view available options
Quantity:
25 g

3-Bromo-4-methoxybenzonitrile is used to prepare 3-bromo-2-iodo-4-methoxy-benzonitrile. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methoxybenzonitrile is used to prepare 3-bromo-2-iodo-4-methoxy-benzonitrile. It is also used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 117572-79-9
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.05
MDL Number MFCD00143097
InChI Key QHWZMDRKTYTPEE-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanoanisole, 3-bromo-4-methoxylbenzonitrile, 3-bromo-4-methoxybenzenecarbonitrile, 3-bromo-4-methoxy-benzonitrile, benzonitrile, 3-bromo-4-methoxy, pubchem3795, acmc-1cexj, maybridge4_001971, 3-bromo4-methoxybenzonitrile, ksc494m8l
PubChem CID 736049
IUPAC Name 3-bromo-4-methoxybenzonitrile
SMILES COC1=CC=C(C=C1Br)C#N

Specifications

Melting Point 120°C to 124°C
Quantity 25 g
UN Number UN3439
Beilstein 3246576
Solubility Information Soluble in methanol.
Formula Weight 212.05
Percent Purity 99%
Chemical Name or Material 3-Bromo-4-methoxybenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.