Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-bromo-1-methyl-1H-pyrazole, 97+%, Thermo Scientific Chemicals

Catalog No. AAB2159206
Click to view available options
Quantity:
0.25g
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146941-72-2
Molecular Formula C4H6BrN3
Molecular Weight (g/mol) 176.02
MDL Number MFCD00663065
InChI Key WYPUMGAZSVZUGJ-UHFFFAOYSA-N
PubChem CID 2735283
SMILES CN1C=C(Br)C(N)=N1

Specifications

Melting Point 96°C to 100°C
Quantity 5 g
Synonym 4-bromo-1-methyl-1h-pyrazol-3-amine, 3-amino-4-bromo-1-methyl-1h-pyrazole, 3-amino-4-bromo-1-methylpyrazole, 1h-pyrazol-3-amine, 4-bromo-1-methyl, 4-bromo-1-methylpyrazole-3-ylamine, acmc-1bzbh, 1h-pyrazol-3-amine,4-bromo-1-methyl
IUPAC Name 4-bromo-1-methylpyrazol-3-amine
Formula Weight 176.02
Percent Purity ≥97%
Chemical Name or Material 3-Amino-4-bromo-1-methyl-1H-pyrazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.