Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3' - Diaminobenzidine Tetrahydrochloride White to pink, crystalline powder MP Biomedicals

Catalog No. 0215082501
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 7411-49-6
Molecular Formula C12H18Cl4N4
Molecular Weight (g/mol) 360.10
MDL Number MFCD08273058,MFCD00012969
InChI Key KJDSORYAHBAGPP-UHFFFAOYSA-N
Synonym DAB, 3, 3', 4
IUPAC Name tetrahydrogen [1,1'-biphenyl]-3,3',4,4'-tetramine tetrachloride
SMILES [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NC1=CC=C(C=C1N)C1=CC=C(N)C(N)=C1

Specifications

Quantity 1 g
Formula Weight 360.1
Physical Form Crystalline Powder
Chemical Name or Material 3, 3' - Diaminobenzidine Tetrahydrochloride
Recommended Storage Store at 0°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.