Learn More
3,3-Diaminobenzidine Tetrahydrochloride, MP Biomedicals™
Click to view available options
Quantity:
5 g
10 g
Description
- 3,3-Diaminobenzidine Tetrahydrochloride is a reagent used in colorimetric analysis of Se and other metals.
- DAB is utilized in many applications for visualization of peroxidase activity.
- In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide; the oxidized DAB forms an insoluble brown end-product.
Specifications
Specifications
Form | Powder |
Quantity | 10 g |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.