Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,3′,3′-Tetramethyl-1,1′-bis(4-sulfobutyl)benzoindodicarbocyanine Sodium Salt 98.0+%, TCI America™
SDP

Catalog No. D44635G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 64285-36-5
Molecular Formula C41H47N2NaO6S2+
Molecular Weight (g/mol) 750.945
MDL Number MFCD16621036
InChI Key ZOYVHUDSHDNKLZ-UHFFFAOYSA-O
Synonym Sodium 4-[2-[5-[1,1-Dimethyl-3-(4-sulfonatobutyl)-1H-benz[e]indol-2(3H)-ylidene]-1,3-pentadienyl]-1,1-dimethyl-1H-benz[e]indol-3-ium-3-yl]butane-1-sulfonate, 2-[5-[1,1-Dimethyl-3-(4-sulfobutyl)-1,3-dihydrobenzo[e]indol-2-ylidene]-1,3-pentadienyl]-1,1-dime
PubChem CID 129894317
IUPAC Name 4-[(2E)-2-[(2E,4E)-5-[1,1-dimethyl-3-(4-sulfobutyl)benzo[e]indol-3-ium-2-yl]penta-2,4-dienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonic acid;sodium
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)O)C=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)O)C=CC6=CC=CC=C65)(C)C)C.[Na]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 748.93
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,3′,3′-Tetramethyl-1,1′-bis(4-sulfobutyl)benzoindodicarbocyanine Sodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.