Learn More
Sklar Instruments Scissors, Sklar Extra Delicate Iris Scissors, Sklar -1/2 in., Sharp/Sharp, Iris Scissors, Curved, Smooth, EA.
Supplier: Sklar Instruments 231154
Scissors, Sklarlite Extra Delicate Iris Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Iris Scissors, Curved, Smooth, EA. Sklarlite XD Iris Scissors are noticeably lighter and delicate than standard Iris Scissors which allows the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. Originally designed for ophthalmic procedures, iris scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp, finger ring scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.