Learn More
Enzo Life Sciences (2R,4R)-APDC (5mg). CAS: 169209-63-6
SDP
Supplier:  Enzo Life Sciences ALX550178M005
Encompass_Preferred
Selective agonist for group II mGluRs. Inhibits the NADPH-oxidase dependent ROS system. Alternative name: LY-314,593, (2R,4R)-4-Aminopyrrolidine-2,4-dicarboxylic acid. Purity: ≥98% (HPLC). Appearance: Off-white powder. Solubility: Soluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-9493
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.