Learn More
Andwin Scientific .2ML RIGID PCR MICROLTRAPLATES
Supplier: Andwin Scientific 21970
The PCR microplates have 96 rigid wells that are made to remove plate distortion and give an appropriate fitting between the thermal cycler and the plate. This product has thin walls that permit quick uniform heat transfer and grid that allows easy identification of samples. This product comes in a case of 25 autoclavable plates.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.