Learn More
Andwin Scientific 2ML DLPHN MCRCNTRFG TB 1000/PK
SDP
Supplier:  Andwin Scientific C32051
Encompass_Preferred
The Sorenson 2.0mL Dolphin Microcentrifuge Tubes are recommended for pelletizing samples. This product can operate within the temperature range of -80˚C to 121˚C. The tubes are compatible with all standard rotors and will not pop open under pressure when boiling due to its positive seal design. It is sold in a pack of 1000.
Catalog No. 50-999-381
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.