Learn More
Gibco™ 293fectin™ Transfection Reagent
293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells.
Supplier: Gibco™ 12347750
Description
293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells. This reagent is optimized for transfecting suspension 293 human embryonic kidney cells (e.g., FreeStyle™ 293-F cells, Cat. No. R790-07) in defined, serum-free FreeStyle™ 293 Expression Medium (12338-018), and is intended for use with the FreeStyle™ 293 Expression System (K9000-01).
293fectin™ Transfection Reagent provides the following advantages:
- Demonstrates high transfection efficiency in suspension 293 cells and is also suitable for transfecting adherent 293 cells
- Suspension FreeStyle™ 293-F cells may be transfected in FreeStyle™ 293 Expression Medium; no medium change is required
- Add 293fectin™ reagent complexes directly to cells in culture medium
- No need to remove complexes or change or add medium following transfection
Specifications
Each kit contains 10 units. Each unit contains one vial of reagent. Store at 4°C. 293fectin™ is guaranteed stable for 6 months when stored properly. |
|
Tube | |
Lipid-based Transfection | |
Not High-throughput Compatible (Manual) | |
Transfection Reagent | |
Yes |
Established Cell Lines | |
Plasmid DNA | |
Protein Expression and Transfection | |
293fectin™ | |
10 x 15 mL |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.