Learn More
Enzo Life Sciences 27-Hydroxycholesterol (5mg). CAS: 20380-11-4
Supplier: Enzo Life Sciences ENZCHM1020005
27-Hydroxycholesterol (27-HC) is a metabolite resulting from the action of sterol 27-Hydroxylase on cholesterol. It is an endogenous selective estrogen receptor modulator and ligand for the liver X receptor (LXR). 27-Hydroxycholesterol plays a role in Alzheimer's disease, oxidative stress, metabolism, cell-signaling, and heart disease. Alternative name: 5,25R-Cholesten-3β,26-diol, (25R)-26-Hydroxycholesterol. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (up to 10 mg/ml with warming) or in ethanol (up to 10 mg/ml). Insoluble in water. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.