Learn More
MTC Bio 250mL Centrifuge Tube, PP, sterile, with screw-cap, 14 bags of 5 tubes, 70/cs
Supplier: MTC Bio C1250
250mL screw-cap polypropylene tubes are manufactured of polypropylene for temperature and chemical resistance. These tubes are intended for large volume centrifugation and are widely used in cell harvesting/purification and drug discovery applications. MTC Bio's 250mL centrifuge tubes can withstand up to 6,000xg maximum RCF*. They are supplied in sterile, RNase, DNase, and pyrogen-free packs of 5 tubes, 70 tubes per case.. Perfect for Cell harvesting. Molded graduations from 0.5 to 250mL. Sterile, RNase, DNase & Pyrogen free. Leak-proof screw-cap
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.