Learn More
Cytiva (Formerly Pall Lab) AcroPrep 24-well Sterile Filtration Plate-0.65/0.2 m Supor EKV (2/pkg)
Supplier:  Cytiva 97017
Encompass_Preferred
AcroPrep 24-well Sterile Filtration Plate-0.65/0.2 m Supor EKV (2/pkg). Palls sterile filtration AcroPrep 24-well filter plate iswell suited for high volume (up to 7 mL) plate-basedsterile filtration needs such as media, reagent, serumor proteins. Compatible with either a vacuum manifoldor centrifuge, the 24-well Supor EKV plate features adual-layer, 0.65 m membrane integrated with a highlyasymmetric 0.2 m membrane for fast, efficient, andsterilizing grade filtration.
Catalog No. NC1857353
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.