Learn More
Chemglass Life Sciences Glas-col Mantle Extension Support for Series inMin, Mantle Size 2000-3000mL, 259mm ID
Supplier: Chemglass Life Sciences CG1100404
Glas-col Mantle Extension Support for Series inMin, Mantle Size 2000-3000mL, 259mm ID. Glas-Col extension support for use with a smaller Series M mantle. Attached to a stable ring stand or distillation rack, the support can be adjusted to any convenient height. Support is painted gray and has a 6in long, 1/2in O.D. rod.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.