Learn More
Enzo Life Sciences 20-Hydroxyecdysone (10mg). CAS: 5289-74-7
SDP
Supplier:  Enzo Life Sciences ALX370012M010
Encompass_Preferred
Most widely occurring ecdysteroid in both plant and animal species. Controls cell death during metamorphosis of Drosophila melanogaster. Alternative name: beta-Ecdysone, 2beta,3beta,14alpha,20R,22R,25-Hexahydroxy-5beta-cholest-7-en-6-one, Ecdysterone. Purity: ≥95% HPLC. Solubility: Soluble in water, 100% ethanol, methanol, DMSO and acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-8892
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.