Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(Phthalimido)ethanesulfonyl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
2-(Phthalimido)ethanesulfonyl chloride is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Phthalimido)ethanesulfonyl chloride is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-(Phthalimido)ethanesulfonyl chloride is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 156°C to 160°C |
Quantity | 1 g |
UN Number | UN3261 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 273.7 |
Percent Purity | 97% |
Chemical Name or Material | 2-(Phthalimido)ethanesulfonyl chloride |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction