Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenyl phenyl ether, 99%, Thermo Scientific Chemicals

Catalog No. AAA1808830
Click to view available options
Quantity:
250 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2216-12-8
MDL Number MFCD00035744

Specifications

Density 1.23
Boiling Point 184°C (8 mmHg)
Flash Point 181°C (358°F)
Molecular Formula C12H9NO3
Refractive Index 1.6 00
Quantity 250 g
Beilstein 2051914
Synonym 2-nitrodiphenyl ether, 2-nitrophenyl phenyl ether, benzene, 1-nitro-2-phenoxy, o-nitrophenyl phenyl ether, 2-nde, phenyl o-nitrophenyl ether, ether, o-nitrophenyl phenyl, ether, 2-nitrophenyl phenyl, ether, 2-nitrodiphenyl,, ganoderic-acid-g
InChI Key VNHGETRQQSYUGZ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2[N+](=O)[O-]
IUPAC Name 1-nitro-2-phenoxybenzene
Molecular Weight (g/mol) 215.21
PubChem CID 16661
Formula Weight 215.21
Percent Purity 99%
Chemical Name or Material 2-Nitrophenyl phenyl ether
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-684-6
RTECSNumber KO2650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.