Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylvaleryl chloride, 98%, Thermo Scientific Chemicals

Catalog No. AAL0272514
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5238-27-7
Molecular Formula C14H12N2O3S
Molecular Weight (g/mol) 288.32
MDL Number MFCD00013656
InChI Key NADADZQYJBFZGI-FLIBITNWSA-N
Synonym 2-methylvaleryl chloride, 2-methylvaleroyl chloride, 2-methylvalerylchloride, .alpha.-methyl valeroyl chloride, acmc-20anpk, 2-methyvaleryl chloride, pubchem21794, 2-methyl valeryl chloride, 2-methyl-valeryl chloride, 2-methyl pentanoyl chloride
PubChem CID 107385
SMILES CC1=CC=C(C=C1)C(\O)=C\C(=O)C(=O)NC1=NC=CS1

Specifications

Density 0.978
Boiling Point 140°C to 144°C
Flash Point 37°C (98°F)
Odor Acrid
Refractive Index 1.4239
Quantity 25 g
UN Number UN2920
Beilstein 1743993
Sensitivity Moisture sensitive
IUPAC Name 2-methylpentanoyl chloride
Formula Weight 134.61
Percent Purity 98%
Chemical Name or Material 2-Methylvaleryl chloride
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 226-035-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.