Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylsulphonylbenzoic acid, 95%, Thermo Scientific™

Catalog No. AC424830025
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 33963-55-2
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.22
MDL Number MFCD00216492
InChI Key BZSXEZOLBIJVQK-UHFFFAOYSA-N
Synonym 2-methylsulfonyl benzoic acid, 2-methanesulfonylbenzoic acid, 2-methanesulfonyl-benzoic acid, 2-methanesulfonyl benzoic acid, benzoic acid, 2-methylsulfonyl, 2-methylsulfonyl benzenecarboxylic acid, 2-methylsulphonyl benzoic acid, 2-mesylbenzoic acid, pubchem18527, methylsulfonylbenzoic acid
PubChem CID 284342
IUPAC Name 2-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=CC=C1C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 136.0°C to 141.0°C
Color Green or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula SO2CH3C6H4COOH
Quantity 2.5 g
Solubility Information Solubility in water: negligible
Formula Weight 200.22
Percent Purity 95%
Physical Form Chunks and/or Powder
Chemical Name or Material 2-Methylsulphonylbenzoic acid, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.